Order Entry
Export
ContactUsLinkComponent
2,2-Diethoxypropane, Sigma-Aldrich®
2,2-Diethoxypropane, Sigma-Aldrich®
Catalog # SIAL519618-25G
Supplier:  Merck
CAS Number:  
2,2-Diethoxypropane, Sigma-Aldrich®
Catalog # SIAL519618-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :132.2 g/mol
Boiling Pt:61 °C (1013 hPa)
Density:0.826 g/cm³ (18 °C)
ADR:3,II
 :M-Clarity™ quality level = MQ100
EINECS:204-808-6
UN:3271
CAS Number:126-84-1
Formula:C₇H₁₆O₂
MDL Number:MFCD00009224