

9-(2,6-Dimethylphenyl)-10-methylacridinium perchlorate
Catalog # TCIAD3429-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
Specification Test Results
About this item
:
:397.86 g/mol
UN:1479
ADR:5.1,III
:GHS03
:GHS07
CAS Number:1276539-32-2
Formula:C₂₂H₂₀ClNO₄