2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10191-18-1
:
Formula:C₆H₁₅NO₅S
MDL Number:MFCD00007533
Boiling Pt:405 °C (1013 hPa)
Melting Pt:152…156 °C
:M-Clarity™ quality level = MQ400
:213.25 g/mol
Storage Temperature:Ambient
EINECS:233-465-5