Order Entry
Export
ContactUsLinkComponent
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0% (by GC)
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0% (by GC)
  TCIAD2802-200MG
 :  TCI
 :  
(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0% (by GC)
  TCIAD2802-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    180.0 to 185.0 deg-C
  • Specific rotation [a]20/D:
    -10.0 to -14.0 deg (C=1, Chloroform)
  • Solubility in Chloroform:
    almost transparency

 

 

 :
 :472.18 g/mol
 :75714-60-2
 :C₂₂H₁₆Br₂O₂
 :MFCD04038415