
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10191-18-1
:
Formula:C₆H₁₅NO₅S
MDL Number:MFCD00007533
Boiling Pt:405 °C (1013 hPa)
Melting Pt:152…154 °C
:213.25 g/mol
Storage Temperature:Ambient
EINECS:233-465-5
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (100°C, 2 hrs)
- Sulfated ash:≤0.1 %
- Solubility:(1 M in water) Clear colorless
- UV:(1 M in water)
- Titration with CHA:≥99.0 %
- Heavy metals (as Pb):≤5 ppm