Order Entry
Export
ContactUsLinkComponent
Ethyl-cis-4,7,10,13,16,19-docosahexaenoate ≥97.0% (by GC)
Ethyl-cis-4,7,10,13,16,19-docosahexaenoate ≥97.0% (by GC)
Catalog # TCIAD2195-100MG
Supplier:  TCI
CAS Number:  
Ethyl-cis-4,7,10,13,16,19-docosahexaenoate ≥97.0% (by GC)
Catalog # TCIAD2195-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :356.55 g/mol
Storage Temperature:Refrigerator
CAS Number:84494-72-4
Formula:C₂₄H₃₆O₂
MDL Number:MFCD00056293