
1,3-Diiodopropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:627-31-6
:
Formula:C₃H₆I₂
MDL Number:MFCD00001097
:295.89 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:–20 °C
Density:2.59617 g/cm³ (4 °C)
:M-Clarity™ quality level = MQ200
EINECS:210-993-4
:GHS07