
S-[2-(Dimethylamino)ethyl]isothiourea dihydrochloride ≥97.0% (by titrimetric analysis)
Catalog # TCIAD0652-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:179.0 to 183.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:220.16 g/mol
EINECS:240-281-9
:GHS07
CAS Number:16111-27-6
Formula:C₅H₁₃N₃S
MDL Number:MFCD00060161