
Silver diethyldithiocarbamate analytical reagent
Catalog # 21563.108
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:256.14 g/mol
Melting Pt:176…178,5 °C
Storage Temperature:Refrigerator
EINECS:216-003-7
UN:3077
ADR:9,III
:GHS07
CAS Number:1470-61-7
Formula:(C₂H₅)₂NCSSAg
MDL Number:MFCD00004929