Order Entry
Export
ContactUsLinkComponent
Silver diethyldithiocarbamate 99% for analysis
Irritant
Silver diethyldithiocarbamate 99% for analysis
Certificates
undefined
Irritant
Silver diethyldithiocarbamate 99% for analysis

Specification Test Results

  • Appearance:
    Green to khaki Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.5% in pyridine) clear
  • Titration with NH4SCN:
    ≥98.5 %

Specification Test Results

Related Information