Order Entry
Export
ContactUsLinkComponent
1-Cyclopentylpiperazine ≥98.0% (by GC, titration analysis)
Irritant
1-Cyclopentylpiperazine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC2466-5G
Supplier:  TCI
CAS Number:  
Irritant
1-Cyclopentylpiperazine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC2466-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :154.26 g/mol
Boiling Pt:105°C
 :GHS07
CAS Number:21043-40-3
Formula:C₉H₁₈N₂
MDL Number:MFCD00168029