Order Entry
Export
ContactUsLinkComponent
2-(4-Chlorophenyl)benzothiazole ≥98.0% (by GC)
2-(4-Chlorophenyl)benzothiazole ≥98.0% (by GC)
Catalog # TCIAC2002-5G
Supplier:  TCI
CAS Number:  
2-(4-Chlorophenyl)benzothiazole ≥98.0% (by GC)
Catalog # TCIAC2002-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :245.72 g/mol
CAS Number:6265-91-4
Formula:C₁₃H₈ClNS
MDL Number:MFCD00030229