Order Entry
Export
ContactUsLinkComponent
N-Cyclohexylformamide ≥98.0% (by GC)
N-Cyclohexylformamide ≥98.0% (by GC)
  TCIAC1436-25G
 :  TCI
 :  
N-Cyclohexylformamide ≥98.0% (by GC)
  TCIAC1436-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :127.19 g/mol
 :151°C
 :766-93-8
 :C₇H₁₃NO
 :MFCD00003828