
Calcium acetylacetonate hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0930-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh substance)
- Water:max. 15.0 %
Specification Test Results
About this item
:
:238.30 g/mol (anhy)
EINECS:243-001-3
:GHS07
CAS Number:19372-44-2
Formula:C₁₀H₁₄CaO₄·xH₂O
MDL Number:MFCD00013486