
Isopropyl hexanoate ≥98.0% (by GC)
TCIAC0841-25ML
: TCI
:
- Pk:25 mL
- Seq:2315
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
:
:168°C
:54°C
:3272
:3,III
:158.24 g/mol
:219-000-9
:GHS02
:2311-46-8
:C₉H₁₈O₂
:MFCD00059436