
Cinchonine sulphate dihydrate ≥98.0% (by titrimetric analysis)
TCIAC0791-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 85.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+188 to +192 deg (C=1, EtOH)
- Water:4.7 to 5.2 %
:
:686.87 g/mol
:227-708-4
:GHS07
:5949-16-6
:C₁₉H₂₂N₂O
:MFCD00035639