
Cinchonidine sulphate dihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0349-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:6.5 to 7.2 %
Specification Test Results
About this item
:
:490.54 g/mol
EINECS:208-362-3
:GHS07
CAS Number:524-61-8
Formula:C₁₉H₂₂N₂O
MDL Number:MFCD00067565