Cholesteryl stearate ≥93.0% (by titrimetric analysis)
Catalog # TCIAC0323-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Ester Value):93.0 to 107.0 %
- Specific rotation [a]20/D:-21.0 to -25.0 deg (C=2, CHCl3)
Specification Test Results
About this item
:
:653.13 g/mol
EINECS:252-637-0
CAS Number:35602-69-8
Formula:C₄₅H₈₀O₂
MDL Number:MFCD00003639