
Decanoic anhydride ≥98.0% (by GC)
Catalog # TCIAC0020-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Morpholine Method):min. 97.0 %
- Solubility in Toluene:almost transparency
Specification Test Results
About this item
:
ADR:8,III
:326.52 g/mol
EINECS:218-213-4
UN:3261
:GHS05
Storage Temperature:Refrigerator
CAS Number:2082-76-0
Formula:C₂₀H₃₈O₃
MDL Number:MFCD00010460