


(±)-Camphene ≥78.0% (by GC) (contains ca. 20% Tricyclene)
Catalog # TCIAC0009-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 78.0 %
- Melting point:44.0 to 48.0 deg-C
Specification Test Results
About this item
:
:136.24 g/mol
Merck Index:14,01730
Boiling Pt:159°C
EINECS:201-234-8
UN:1325
ADR:4.1,II
:GHS02
:GHS07
:GHS09
CAS Number:79-92-5
Formula:C₁₀H₁₆
MDL Number:MFCD00066603