
Cacotheline monohydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:561-20-6
:
Formula:C₂₁H₂₁N₃O₇
MDL Number:MFCD00077211
:490.43 g/mol
Merck Index:14,01605
EINECS:209-215-6
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %