2-Ethoxyethyl acetate
Glass bottle for solids, 1 mL
Catalog # EHERC10016000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
Synonyms: Ethylglycol acetate
- CAS Number:111-15-9
- Formula:C₆H₁₂O₃
- MDL Number:MFCD00009251
- Molecular Weight:132.16 g/mol
- Merck Index:13,03787
- Boiling Pt:156 °C (1013 hPa)
- Melting Pt:–62 °C
- Density:0.976 g/cm³ (20 °C)
- Flash Pt:54 °C
- EINECS:203-839-2
- UN:1172
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mL



