2-Ethoxyethyl acetate ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 111-15-9 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 52°(126°F) Refractive Index: 1.4070 Odour: Fruit-like Melting Point: -62° Boiling Point: 155-157° Beilstein Reference: 1748677 Merck Reference: 14,3751 UN No.: 1172 MDL No.: MFCD00009251
Synonyms: Ethylglycol acetate
- CAS Number:111-15-9
- Formula:C₆H₁₂O₃
- MDL Number:MFCD00009251
- Molecular Weight:132.16 g/mol
- Merck Index:14,3751
- Boiling Pt:155…157 °C
- Melting Pt:–62 °C
- Density:0.975
- Flash Pt:52 °C (126 °F)
- Storage Temperature:Ambient
- EINECS:203-839-2
- UN:1172
- ADR:3,III
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 111-15-9 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 52°(126°F) Refractive Index: 1.4070 Odour: Fruit-like Melting Point: -62° Boiling Point: 155-157° Beilstein Reference: 1748677 Merck Reference: 14,3751 UN No.: 1172 MDL No.: MFCD00009251
Synonyms: Ethylglycol acetate
- CAS Number:111-15-9
- Formula:C₆H₁₂O₃
- MDL Number:MFCD00009251
- Molecular Weight:132.16 g/mol
- Merck Index:14,3751
- Boiling Pt:155…157 °C
- Melting Pt:–62 °C
- Density:0.975
- Flash Pt:52 °C (126 °F)
- Storage Temperature:Ambient
- EINECS:203-839-2
- UN:1172
- ADR:3,III



