Methyl myristate
Glass bottle for solids, 100 mg
Catalog # ACCUSFA007N
Supplier: AccuStandard
Jump to:
Product Details & Documents
Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate
- CAS Number:124-10-7
- Formula:C₁₅H₃₀O₂
- MDL Number:MFCD00008983
- Molecular Weight:242.4 g/mol
- Boiling Pt:323 °C (1013 hPa)
- Melting Pt:18 °C
- Density:0.86715 g/cm³ (20 °C)
- Flash Pt:140 °C
- Storage Temperature:Refrigerator
- EINECS:204-680-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



