Methyl palmitate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-39-0
:
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008994
:270.46 g/mol
Boiling Pt:417 °C (1013 hPa)
Melting Pt:29.9 °C
Density:0.8804 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:203-966-3