Order Entry
Export
ContactUsLinkComponent
 
Methyl palmitate
Catalog # EHERC14192100
Supplier:  EHRENSTORFER
CAS Number:  
Methyl palmitate
Catalog # EHERC14192100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :270.46 g/mol
Boiling Pt:417 °C (1013 hPa)
Melting Pt:29.9 °C
Density:0.8804 g/cm³ (20 °C)
EINECS:203-966-3
CAS Number:112-39-0
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008994