Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl palmitate
Methyl palmitate
Methyl palmitate
Methyl palmitate
Supplier:
Cayman Chemical
Certificates
Expand image
Methyl palmitate
Supplier:
Cayman Chemical
Certificates
About this item
CAS Number:
112-39-0
:
Formula:
C₁₇H₃₄O₂
MDL Number:
MFCD00008994
:
270.46 g/mol
Boiling Pt:
417 °C (1013 hPa)
Melting Pt:
29.9 °C
Density:
0.8804 g/cm³ (20 °C)
EINECS:
203-966-3
Related Information
Documentation
Certificates
Search by Lot Number
Documentation