Order Entry
Export
ContactUsLinkComponent
tert-Butyl glycidyl ether ≥96.0% (by GC)
Flammable
Irritant
tert-Butyl glycidyl ether ≥96.0% (by GC)
Catalog # TCIAB1840-25ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
tert-Butyl glycidyl ether ≥96.0% (by GC)
Catalog # TCIAB1840-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :130.19 g/mol
Boiling Pt:152°C
Flash Pt:44°C
EINECS:231-640-0
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:7665-72-7
Formula:C₇H₁₄O₂
MDL Number:MFCD00010050