2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥98.0% (by HPLC)
Catalog # TCIAB1767-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Melting point:136.0 to 138.0 deg-C
Specification Test Results
About this item
:
:354.45 g/mol
EINECS:239-135-7
CAS Number:15082-28-7
Formula:C₂₄H₂₂N₂O
MDL Number:MFCD00003101