
4,4'-Biphenyldisulphonic Acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1664-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:314.33 g/mol
UN:2585
ADR:8,III
:GHS05
CAS Number:5314-37-4
Formula:C₁₂H₁₀O₆S₂
MDL Number:MFCD00035762