
5-tert-Butylisophthalic acid ≥99.0% (by GC, titration analysis)
Catalog # TCIAB1380-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:222.24 g/mol
EINECS:219-100-2
:GHS07
CAS Number:2359-09-3
Formula:C₁₂H₁₄O₄
MDL Number:MFCD00067047