Order Entry
Export
ContactUsLinkComponent
Triacetin (glycerol triacetate)
Triacetin (glycerol triacetate)
Catalog # MOLE27238415-1KG
Supplier:  Molekula
CAS Number:  
Triacetin (glycerol triacetate)
Catalog # MOLE27238415-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1.1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716