Order Entry
Export
ContactUsLinkComponent
2,3-Butanedione monoxime analytical reagent
2,3-Butanedione monoxime analytical reagent
Supplier:  Merck
Certificates
2,3-Butanedione monoxime analytical reagent
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    74 - 76 °C
  • Molar Mass:
    101.1 g/mol
  • Bulk Density:
    440 kg/m3
  • Boiling Point:
    185 - 186 °C

Specification Test Results

Related Information