Trichloroacetyl isocyanate
Catalog # MOLEM66744723
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:188.4 g/mol
Boiling Pt:80…85 °C (27 hPa)
Density:1.59 g/cm³ (20 °C)
Flash Pt:65 °C
EINECS:221-165-7
UN:2927
ADR:6.1,II
CAS Number:3019-71-4
Formula:C₃Cl₃NO₂
MDL Number:MFCD00002033