


Trichloroacetyl isocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3019-71-4
:
Formula:C₃Cl₃NO₂
MDL Number:MFCD00002033
:188.4 g/mol
Boiling Pt:58…60 °C (20 mmHg)
Density:1.585
Flash Pt:65 °C (149 °F)
EINECS:221-165-7
UN:2927
ADR:6.1,II
:GHS06
:GHS08
:GHS05