Order Entry
Export
ContactUsLinkComponent
Ruthenium(III) chloride hydrate
Corrosive
Irritant
Ruthenium(III) chloride hydrate
Catalog # ROTH7910.1
Supplier:  Roth Carl
CAS Number:  
Corrosive
Irritant
Ruthenium(III) chloride hydrate
Catalog # ROTH7910.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :207.43 g/mol (anhy)
Melting Pt:> 300 °C
Density:3.11 g/cm³ (20 °C)
EINECS:233-167-5
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:14898-67-0
 :20759-14-2
Formula:Cl₃Ru·xH₂O
MDL Number:MFCD00149844