L(+)-Lysine monohydrate
Catalog # ROTH4207.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
Melting Pt:225 °C
:164.2 g/mol
Storage Temperature:Refrigerator
EINECS:200-294-2
CAS Number:39665-12-8
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MDL Number:MFCD00151035