L(+)-Lysine monohydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39665-12-8
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MDL Number:MFCD00151035
Melting Pt:212…214 °C (dec.)
:164.2 g/mol
Storage Temperature:Ambient
EINECS:200-294-2
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:9.5 to 11.5 % (K.F.)
- Sulfated ash:≤0.1 %
- pH:9.0 to 10.5 (2.5% in H2O, 25°C)
- Specific optical rotation:+22.0° to +27.0° (20°C, 589 nm) (c=8, 6 N HCl) (On dry basis)
- Titration with HClO4:98.5 to 101.5 %
- Chloride (Cl):≤300 ppm
- Sulfate (SO4):≤200 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Other amino acids:≤0.5 % (TLC)