Order Entry
Export
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
Irritant
Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
Catalog # SIALM1019-100G
Supplier:  Merck
CAS Number:  
Irritant
Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
Catalog # SIALM1019-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :1235.86 g/mol
Merck Index:13,00536
Density:2.498 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:234-722-4
 :GHS07
Storage Temperature:Ambient
CAS Number:12054-85-2
 :12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059