Order Entry
Export
Orders LinkContactUsLinkComponent
 

Indium(III) nitrate hydrate, Sigma-Aldrich®

Supplier:   Merck
Indium(III) nitrate hydrate, Sigma-Aldrich®
Supplier:  Merck
Certificates
Indium(III) nitrate hydrate, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Indium trinitrate hydrate
  • CAS Number:207398-97-8
  • Formula:InN₃O₉·H₂O
  • MDL Number:MFCD00149742
  • Molecular Weight:318.85 g/mol
  • ADR:5.1,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:237-393-5
  • UN:1477
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 126,00 €
    126,00 €
    1 * 10 g
       126,00 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 498,00 €
    498,00 €
    1 * 50 g
       498,00 €