Order Entry
Export
ContactUsLinkComponent
Sodium sulphide hydrate
Toxic
Corrosive
Environment
Sodium sulphide hydrate
Catalog # ROTH8634.1
Supplier:  Roth Carl
CAS Number:  
Toxic
Corrosive
Environment
Sodium sulphide hydrate
Catalog # ROTH8634.1
Supplier:  Roth Carl
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

About this item

 :
 :96.06 g/mol (anhy)
Merck Index:12,08829
Boiling Pt:∼920 °C (1013 hPa)
Melting Pt:50 °C
Density:∼1.43 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:215-211-5
UN:1849
ADR:8,II
REACH:01-2119513694-38
 :GHS06
 :GHS05
 :GHS09
CAS Number:27610-45-3
Formula:Na₂S·xH₂O
MDL Number:MFCD00149183