Order Entry
Export
Orders LinkContactUsLinkComponent
L(-)-Cystine

L(-)-Cystine

Glass bottle for solids, 2 g
Catalog # PROONIST-143D
L(-)-Cystine
Catalog # PROONIST-143D
CAS Number:  56-89-3
L(-)-Cystine

Glass bottle for solids, 2 g

CAS Number:  56-89-3
Price
887,00 €
Log in for availability
1 * 2 g
Product Details & Documents
Synonyms:  (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
  • CAS Number:56-89-3
  • Formula:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
  • MDL Number:MFCD00064228
  • Molecular Weight:240.3 g/mol
  • Merck Index:13,02811
  • Boiling Pt:428 °C (1013 hPa)
  • Melting Pt:261…262 °C
  • Density:1.677 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-296-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g

Specifications