L(-)-Cystine
Glass bottle for solids, 100 g
Catalog # MOLE36410013-100G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
- CAS Number:56-89-3
- Formula:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
- MDL Number:MFCD00064228
- Molecular Weight:240.3 g/mol
- Merck Index:13,02811
- Boiling Pt:428 °C (1013 hPa)
- Melting Pt:261…262 °C
- Density:1.677 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-296-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



