L(-)-Cystine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 56-89-3 Molecular Formula: C6H12N2O4S2 Formula Weight: 240.30 Physical Form: Cryst powder Density: 1.68 Appearance: White Melting Point: 240° dec. Beilstein Reference: 1728094 Merck Reference: 14,2782 Optical Rotation: -219° (c=1 in 1N HCl) MDL No.: MFCD00064228
Synonyms: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
- CAS Number:56-89-3
- Formula:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
- MDL Number:MFCD00064228
- Molecular Weight:240.3 g/mol
- Merck Index:14,2782
- Boiling Pt:428 °C (1013 hPa)
- Melting Pt:>240 °C (dec.)
- Density:1.68
- Storage Temperature:Ambient
- EINECS:200-296-3
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 56-89-3 Molecular Formula: C6H12N2O4S2 Formula Weight: 240.30 Physical Form: Cryst powder Density: 1.68 Appearance: White Melting Point: 240° dec. Beilstein Reference: 1728094 Merck Reference: 14,2782 Optical Rotation: -219° (c=1 in 1N HCl) MDL No.: MFCD00064228
Synonyms: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
- CAS Number:56-89-3
- Formula:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
- MDL Number:MFCD00064228
- Molecular Weight:240.3 g/mol
- Merck Index:14,2782
- Boiling Pt:428 °C (1013 hPa)
- Melting Pt:>240 °C (dec.)
- Density:1.68
- Storage Temperature:Ambient
- EINECS:200-296-3



