Sodium hypophosphite monohydrate, (H₂O 12-17%) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10039-56-2
:
Formula:NaH₂PO₂·H₂O
MDL Number:MFCD06252379
:105.99 g/mol
Merck Index:14,8629
Melting Pt:90 °C
Density:0.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-669-9