Order Entry
Export
ContactUsLinkComponent
Sodium molybdate(VI) dihydrate, Sigma-Aldrich®
Sodium molybdate(VI) dihydrate, Sigma-Aldrich®
Catalog # SIALM1651-100G
Supplier:  Merck
CAS Number:  
Sodium molybdate(VI) dihydrate, Sigma-Aldrich®
Catalog # SIALM1651-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-551-7
CAS Number:10102-40-6
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170