


Hexachloroplatinic(IV) acid hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:26023-84-7
:
Formula:H₂Cl₆Pt·H₂O
MDL Number:MFCD00149909
:427.83 g/mol
Melting Pt:60 °C
EINECS:241-010-7
UN:2507
ADR:8,III
:GHS06
:GHS05
:GHS08