Order Entry
Export
ContactUsLinkComponent
Phosphotungstic acid hydrate, AnalaR NORMAPUR® analytical reagent sodium-free
Corrosive
Phosphotungstic acid hydrate, AnalaR NORMAPUR® analytical reagent sodium-free
Catalog # 20636.183
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Phosphotungstic acid hydrate, AnalaR NORMAPUR® analytical reagent sodium-free
Catalog # 20636.183
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Loss on ignition (750°C):
    Max. 17 %
  • Total N (Nitrogen):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 20 ppm
  • K (Potassium):
    Max. 0.02 %
  • Na (Sodium):
    Max. 0.02 %
  • Pb (Lead):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :2898.07 g/mol
Merck Index:13,07450
Melting Pt:107 °C
EINECS:215-682-7
UN:3260
ADR:8,III
 :GHS05
CAS Number:12501-23-4
 :12067-99-1
Formula:H₃[P(W₃O₁₀)₄]·nH₂O
MDL Number:MFCD00011342