Order Entry
Export
ContactUsLinkComponent
Dodecamolybdophosphoric acid hydrate
Oxidizing
Corrosive
Dodecamolybdophosphoric acid hydrate
Catalog # ROTH4440.1
Supplier:  Roth Carl
CAS Number:  
Oxidizing
Corrosive
Dodecamolybdophosphoric acid hydrate
Catalog # ROTH4440.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :1843.27 g/mol
Merck Index:12,07498
Melting Pt:78…90 °C
Density:1.62 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-713-5
UN:3260
ADR:8,III
 :GHS03
 :GHS05
CAS Number:51429-74-4
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913