(S)-(+)-3-Chloro-1,2-propanediol, Sigma-Aldrich®
Catalog # SIAL540064-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:110.54 g/mol
Boiling Pt:80 °C (0.5 mmHg)
Density:1.322 g/cm³ (20 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:2689
CAS Number:60827-45-4
Formula:ClCH₂CHOHCH₂OH
MDL Number:MFCD00210270